Learn More
Enzo Life Sciences DPPIV drug discovery kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences BMLAK4990001
Encompass_Preferred
The DPPIV Drug Discovery Kit is a complete assay system designed to screen DPPIV inhibitors, providing enough material to perform at least 96 assays. DPPIV (DPP4, CD26) is a member of the class of proteases known as prolyl peptidases, which cleave proteins after proline residues and is thought to play roles in diabetes, cancer, and autoimmune diseases, making it a target for drug discovery. The kit contains both a chromogenic substrate (H-Gly-Pro-pNA; Km=114 µM) and a fluorogenic substrate (H-Gly-Pro-AMC; Km=50 µM). Cleavage of the p-nitroaniline (pNA) from the colorimetric substrate increases absorbance at 405 nm. The fluorimetric assay is based on the cleavage of 7-amino-4-methylcoumarin (AMC) moiety from the C-terminus of the peptide substrate, which increases its fluorescence intensity at 460 nm. Alternative name: CD26, Dipeptidyl Peptidase IV, DPP4. Applications: Colorimetric detection, Fluorescent detection, HTS Activity assay.
Catalog No. 50-201-0283