Learn More
Enzo Life Sciences DPPIV (human), (recombinant) (25 mU)
SDP
Supplier:  Enzo Life Sciences BMLSE4340025
Encompass_Preferred
Alternative name: Dipeptidyl Peptidase 4, DPP4, CD26. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 10 mM TRIS, pH 8.0, 100mM NaCl. Source: Produced in Sf9 insect cells using baculovirus expression system. Active recombinant human DPPIV (aa 29-766). This represents a naturally-occurring cleaved (soluble) form of DPPIV. Long Term Storage: -80°C.
Catalog No. 50-201-2023