Learn More
ACROBiosystems Recombinant Protein;Human DPPIV / CD26 Protein, Fc Tag;HEK293;50UG;DP4,DPP4,DPP-4,ADABP,ADCP2,ADCP-2,CD26,CD-26,DPPIV,DPPⅣ,TP103,TP-103
Catalog No. 501657544
Encompass_Preferred
Recombinant Protein;Human DPPIV / CD26 Protein, Fc Tag;50UG;Human DPPIV, Fc Tag (DP4-H5266) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession # NP_001926.2).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 112 kDa. The protein migrates as 90-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7544 Supplier ACROBiosystems Supplier No. DP4H526650UG
Recombinant Protein;Human DPPIV / CD26 Protein, Fc Tag;50UG;Human DPPIV, Fc Tag (DP4-H5266) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession # NP_001926.2).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 112 kDa. The protein migrates as 90-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.