Learn More
ACROBiosystems Recombinant Protein;Human DPPIV / CD26 Protein;HEK293;50UG;DP4,DPP4,DPP-4,ADABP,ADCP2,ADCP-2,CD26,CD-26,DPPIV,DPPⅣ,TP103,TP-103
Catalog No. 501657548
Encompass_Preferred
Recombinant Protein;Human DPPIV / CD26 Protein;50UG;Human DPPIV, His Tag (DP4-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 34 - Pro 766 (Accession # NP_001926.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.4 kDa. The protein migrates as 95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7548 Supplier ACROBiosystems Supplier No. DP4H522150UG
Recombinant Protein;Human DPPIV / CD26 Protein;50UG;Human DPPIV, His Tag (DP4-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 34 - Pro 766 (Accession # NP_001926.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.4 kDa. The protein migrates as 95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.