Learn More
ACROBiosystems Recombinant Protein;Human DPPIV / CD26 Protein, Tag Free;HEK293;50UG;DP4,DPP4,DPP-4,ADABP,ADCP2,ADCP-2,CD26,CD-26,DPPIV,DPPⅣ,TP103,TP-103
Supplier:  ACROBiosystems DP4H521150UG
Encompass_Preferred
Recombinant Protein;Human DPPIV / CD26 Protein, Tag Free;50UG;Human DPPIV, Tag Free (DP4-H5211) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession # NP_001926.2).;This protein carries no "tag". The protein has a calculated MW of 85.4 kDa. The protein migrates as 95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7550