Learn More
Medchemexpress LLC DPQ 10mM 1mL | 129075-73-6 | 302.41 g/mol | C18H26N2O2 | 1 ML
SDP
Catalog No. 5000226835
Encompass_Preferred
DPQ is a selective PARP-1 inhibitor supplied as a ready-to-use 10 mM solution in DMSO for research use. It blocks PARP-1-mediated DNA damage repair and NAD+/ATP consumption, and is used in biochemical and cell-based assays to study PARP-related pathways.
  • Ready-to-use 10 mM solution in DMSO.
  • Pack size 1 mL.
  • High purity (99.68%).
  • Molecular weight 302.41 g/mol.
  • Suitable for biochemical and cell-based assays investigating DNA repair and inflammatory signaling.
Catalog No. 50-002-26835 Supplier Medchemexpress LLC Supplier No. HY11486910MM 1ML
DPQ is a selective PARP-1 inhibitor supplied as a ready-to-use 10 mM solution in DMSO for research use. It blocks PARP-1-mediated DNA damage repair and NAD+/ATP consumption, and is used in biochemical and cell-based assays to study PARP-related pathways.
  • Ready-to-use 10 mM solution in DMSO.
  • Pack size 1 mL.
  • High purity (99.68%).
  • Molecular weight 302.41 g/mol.
  • Suitable for biochemical and cell-based assays investigating DNA repair and inflammatory signaling.