Learn More
Enzo Life Sciences DPQ (5mg). CAS: 129075-73-6
SDP
Supplier:  Enzo Life Sciences ALX270221M005
Encompass_Preferred
Very potent poly(ADP-ribose) polymerase-1 (PARP-1) inhibitor. Formula: C18H26N2O2. MW: 302.4. Purity: ≥98% (HPLC). Appearance: Off-white to light brown solid. Solubility: Soluble in DMSO; slightly soluble in 100% ethanol; insoluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-8437