Learn More
Abcam DPQ, PARP-1 inhibitor, 1MG
Catalog No. 502706596
Encompass_Preferred
MW 302.4 Da, Purity >99%. Potent, selective PARP-1 inhibitor (IC₅₀ = 37 nM). Inhibits NF-κB signaling. Shows anti-inflammatory effects in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6596 Supplier Abcam Supplier No. AB1414611MG
MW 302.4 Da, Purity >99%. Potent, selective PARP-1 inhibitor (IC₅₀ = 37 nM). Inhibits NF-κB signaling. Shows anti-inflammatory effects in vivo.
The product is subject to the following: Abcam Restricted Use Statement