Learn More
Enzo Life Sciences (D-Pro1,2)-retro-(D-HIV-TAT(48-57)) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLEI3840001
Encompass_Preferred
Useful as a control peptide in studies using D-JNKi1 (BML-EI355). This peptide is the cell transporter, and does not contain the JIP (JNK inhibitor peptide) sequence. Purity: ≥98% (TLC). Formulation: Lyophilized. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-0761