Learn More
CPC Scientific H-Arg-DPro-Lys-Pro-Gln-Gln-DPhe-Phe-DTrp-Leu-Met-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific SUBP006A
Encompass
One of the first effective antagonists of substance P. It also shows agonist activity in several in vitro preparations.ONE-LETTER SEQUENCE: R-DPro-KPQQ-DPhe-F-DTrp-LM-NH2MOLECULAR FORMULA: C72H105N19O13S1MOLECULAR WEIGHT:1476.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [77275-70-8], RESEARCH AREA: NeuropeptidesREFERENCES: K. Folkers et al., Acta Physiol. Scan., 111, 505 (1981)
Catalog No. 50-211-0450