Learn More
Cayman Chemical DProPheArgpNAhydrochlor 100mg
SDP
Catalog No. 502941626
Encompass_Preferred
A colorimetric plasma kallikrein substrate; cleavage of the PFR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of plasma kallikrein activity
Catalog No. 50-294-1626 Supplier Cayman Chemical Supplier No. 27569100 MG
A colorimetric plasma kallikrein substrate; cleavage of the PFR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of plasma kallikrein activity