Learn More
Cayman Chemical D-Pro-Phe-Arg-pNA (hydrochloride), Purity: ≥95%, CAS: 62354-56-7, 50 mg
SDP
Catalog No. 502939067
Encompass_Preferred
A colorimetric plasma kallikrein substrate; cleavage of the PFR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of plasma kallikrein activity
Catalog No. 50-293-9067 Supplier Cayman Chemical Supplier No. 2756950 MG
A colorimetric plasma kallikrein substrate; cleavage of the PFR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of plasma kallikrein activity