Learn More
Cayman Chemical DPTA NONOate, Purity: ≥98%, CAS: 146724-95-0, 100 mg
SDP
Catalog No. 502892482
Encompass_Preferred
A NO donor with a half-life of three hours at 37°C, pH 7.4; liberates two moles of NO per mole of parent compound
Catalog No. 50-289-2482 Supplier Cayman Chemical Supplier No. 82110100 MG
A NO donor with a half-life of three hours at 37°C, pH 7.4; liberates two moles of NO per mole of parent compound