Learn More
Cayman Chemical DPTA Nonate 10mg
SDP
Catalog No. 502896273
Encompass_Preferred
A NO donor with a half-life of three hours at 37°C, pH 7.4; liberates two moles of NO per mole of parent compound
Catalog No. 50-289-6273 Supplier Cayman Chemical Supplier No. 8211010 MG
A NO donor with a half-life of three hours at 37°C, pH 7.4; liberates two moles of NO per mole of parent compound