Learn More
Cayman Chemical DPTA NONOate, Purity: ≥98%, CAS: 146724-95-0, 50 mg
SDP
Catalog No. 502896321
Encompass_Preferred
A NO donor with a half-life of three hours at 37°C, pH 7.4; liberates two moles of NO per mole of parent compound
Catalog No. 50-289-6321 Supplier Cayman Chemical Supplier No. 8211050 MG
A NO donor with a half-life of three hours at 37°C, pH 7.4; liberates two moles of NO per mole of parent compound