Learn More
Enzo Life Sciences DPTA NONOate (5mg). CAS: 146724-95-0
SDP
Supplier:  Enzo Life Sciences ALX4300665005
Encompass_Preferred
Nitric oxide (NO) donor. The half-life time is 5 hours at 22°C in 0.1M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0. Alternative name: Dipropylenetriamine NONOate. Purity: ≥98% (NMR). Appearance: White to off-white solid. Solubility: Highly soluble in water. Long Term Storage: -80°C.
Catalog No. 50-200-9173