Learn More
Kyfora Bio DPTAP 100 MG
Catalog No. 502559837
Encompass_Preferred
DPTAP Chloride (1,2-dipalmitoyl-3-trimethylammonium-propane Chloride) is a cationic lipid and an analogue of DOTAP. It can be used in the formation of liposomes and microbubbles that encapsulate nucleic acids, such as miRNA, as an efficient gene delivery system.Key application(s): Gene delivery, miRNA delivery
Catalog No. 50-255-9837 Supplier Kyfora Bio Supplier No. 14487100
DPTAP Chloride (1,2-dipalmitoyl-3-trimethylammonium-propane Chloride) is a cationic lipid and an analogue of DOTAP. It can be used in the formation of liposomes and microbubbles that encapsulate nucleic acids, such as miRNA, as an efficient gene delivery system.Key application(s): Gene delivery, miRNA delivery