Learn More
Kyfora Bio DPTAP 25 MG
Catalog No. 502559836
Encompass_Preferred
DPTAP Chloride (1,2-dipalmitoyl-3-trimethylammonium-propane Chloride) is a cationic lipid and an analogue of DOTAP. It can be used in the formation of liposomes and microbubbles that encapsulate nucleic acids, such as miRNA, as an efficient gene delivery system.Key application(s): Gene delivery, miRNA delivery
Catalog No. 50-255-9836 Supplier Kyfora Bio Supplier No. 1448725
DPTAP Chloride (1,2-dipalmitoyl-3-trimethylammonium-propane Chloride) is a cationic lipid and an analogue of DOTAP. It can be used in the formation of liposomes and microbubbles that encapsulate nucleic acids, such as miRNA, as an efficient gene delivery system.Key application(s): Gene delivery, miRNA delivery