Learn More
Cayman Chemical DPT (hydrochloride), Purity: ≥98%, CAS: 16382-06-2, 10 mg
SDP
Catalog No. 502951983
Encompass_Preferred
A psychedelic drug of the tryptamine class; inhibits the re-uptake of dopamine, serotonin, and norepinephrine (IC50s = 23, 2.9, and 9.1 μM); intended for forensic and research applications
Catalog No. 50-295-1983 Supplier Cayman Chemical Supplier No. 1155110 MG
A psychedelic drug of the tryptamine class; inhibits the re-uptake of dopamine, serotonin, and norepinephrine (IC50s = 23, 2.9, and 9.1 μM); intended for forensic and research applications