Learn More
TARGETMOL CHEMICALS INC DPTIP 5MG
SDP
Catalog No. 502579692
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DPTIP is an effective inhibitor of neutral sphingomyelinase 2 with an IC50 value of 30 nM. purity: 98%
Catalog No. 50-257-9692 Supplier TARGETMOL CHEMICALS INC Supplier No. T600435MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DPTIP is an effective inhibitor of neutral sphingomyelinase 2 with an IC50 value of 30 nM. purity: 98%