Learn More
TARGETMOL CHEMICALS INC DPTN dihydrochloride 10MG
SDP
Catalog No. 502877841
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. DPTN dihydrochloride is a potent and selective A3AR antagonist with Ki values of 1.65, 9.61 and 8.53 nM in human, mouse and rat, respectively. Purity 100%
Catalog No. 50-287-7841 Supplier TARGETMOL CHEMICALS INC Supplier No. T7206710MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. DPTN dihydrochloride is a potent and selective A3AR antagonist with Ki values of 1.65, 9.61 and 8.53 nM in human, mouse and rat, respectively. Purity 100%