Learn More
Medchemexpress LLC Dptn dihydrochloride | 325767-87-1 | 98.5% | 459.39 g/mol | C22H20Cl2N4OS | 10 MG
SDP
Catalog No. 5000286036
Encompass_Preferred
DPTN dihydrochloride is a research-grade dihydrochloride salt that functions as a potent, selective antagonist of the adenosine A3 receptor (A3AR) in human, mouse, and rat. It is supplied as a solid for in vitro and receptor-binding studies and is intended for research use only.
  • Potent A3AR antagonist with high affinity (hA3 Ki = 1.65 nM; mouse = 9.61 nM; rat = 8.53 nM).
  • High purity suitable for biochemical assays (98.47%).
  • Solid, light yellow to yellow physical form for easy handling.
  • Soluble in DMSO (~5 mg/mL) with ultrasonic warming recommended.
  • Stable when stored sealed at -20°C; in solution store at -80°C for long-term.
Catalog No. 50-002-86036 Supplier Medchemexpress LLC Supplier No. HY15315210MG
DPTN dihydrochloride is a research-grade dihydrochloride salt that functions as a potent, selective antagonist of the adenosine A3 receptor (A3AR) in human, mouse, and rat. It is supplied as a solid for in vitro and receptor-binding studies and is intended for research use only.
  • Potent A3AR antagonist with high affinity (hA3 Ki = 1.65 nM; mouse = 9.61 nM; rat = 8.53 nM).
  • High purity suitable for biochemical assays (98.47%).
  • Solid, light yellow to yellow physical form for easy handling.
  • Soluble in DMSO (~5 mg/mL) with ultrasonic warming recommended.
  • Stable when stored sealed at -20°C; in solution store at -80°C for long-term.