Learn More
Medchemexpress LLC DPTN dihydrochloride | 325767-87-1 | C22H20Cl2N4OS | 5 MG
SDP
Catalog No. 5000372253
Encompass_Preferred
DPTN dihydrochloride is a potent and selective human, mouse, and rat A3AR antagonist. It exhibits Ki values of 1.65 nM for human A3AR, 9.61 nM for mouse A3AR, and 8.53 nM for rat A3AR. This product is for research use only.
  • Potent and selective A3AR antagonist
  • High purity (98.47%)
  • Solid appearance
  • Light yellow to yellow color
  • Store at -20°C, sealed, away from moisture
  • Shipped at room temperature in continental US
Catalog No. 50-003-72253 Supplier Medchemexpress LLC Supplier No. HY1531525MG
DPTN dihydrochloride is a potent and selective human, mouse, and rat A3AR antagonist. It exhibits Ki values of 1.65 nM for human A3AR, 9.61 nM for mouse A3AR, and 8.53 nM for rat A3AR. This product is for research use only.
  • Potent and selective A3AR antagonist
  • High purity (98.47%)
  • Solid appearance
  • Light yellow to yellow color
  • Store at -20°C, sealed, away from moisture
  • Shipped at room temperature in continental US