Learn More
AdipoGen DPVP (Resveratrol Analog) (1 mg)
SDP
Supplier:  AdipoGen AGCR13604M001
Encompass_Preferred
Chemical. CAS: 1462947-40-5 | 223386-36-5. Formula: C15H15NO2. MW: 241.3. Anticancer compound. Shown to inhibit progression of prostate cancer cells. Anti-obesity agent. 3T3-L1 adipocyte differentiation and FAS expression inhibitor.
Catalog No. 50-205-3973