Learn More
Sigma-Aldrich DPX Mountant for histology
Catalog No. 01258850 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-258-850 500 mL
01-259-034 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-850 Supplier Sigma-Aldrich Supplier No. 06522500ML
Add to Cart
Add to Cart
Slide mounting medium
DPX (Distyrene, Plasticizer, and Xylene) is a widely utilized mounting medium for immunohistology, specifically designed to be compatible with various detection methods. Introduced in 1939, DPX has become the routine mountant of choice due to its effectiveness in preserving histological stains and providing clear visualization of tissue samples. This mounting medium is a mixture of distyrene (a polystyrene), a plasticizer (initially tricresyl phosphate, now modified to dibutyl phthalate), and xylene, making it a versatile option for histological applications.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Slide Mounting Media
Reagent Type Mounting Solution
Volume (Metric) 500 mL