Learn More
Aviva Systems Biology Corp Dpy19l1 Antibody - C-terminal region (ARP43555_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43555_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dpy19l1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1100001I19Rik, 6030402K12, AI225861, AW558481ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-079