Learn More
Aviva Systems Biology Corp DPYS Antibody - middle region (ARP41468_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41468_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DPYS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DPYS. Alternate names: DHP, DHPaseApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57870