Learn More
Aviva Systems Biology Corp DPYSL2 Antibody - middle region (ARP51959_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51959_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DPYSL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DPYSL2. Alternate names: CRMP2, DHPRP2, DRP-2, DRP2, N2A3, ULIP2, CRMP-2, ULIP-2ApplicationIP, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60656