Learn More
Aviva Systems Biology Corp DPYSL3 Antibody - middle region (ARP51962_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51962_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DPYSL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DPYSL3. Alternate names: CRMP-4, CRMP4, DRP-3, DRP3, ULIP, LCRMP, ULIP-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60657