Learn More
Hach Company DR6000™ UV VIS Spectrophotometer with RFID Technology
Catalog No. 501997797 Shop All Hach Company Products
Encompass
The DR6000™ is the industry’s most advanced laboratory spectrophotometer for water testing. It offers high-speed wavelength scanning across the UV and Visible Spectra, and comes with over 250 pre-programmed methods including the most common water and environmental testing methods. With optional accessories allowing for high-volume testing via a carousel sample changer, and increased accuracy with a sample delivery system that eliminates optical difference errors, this instrument ensures you are ready to handle your wide-ranging water testing needs. Combining the DR6000 with Hach TNTplus™ test kits provides additional accuracy with guided step-by-step testing procedures, auto-method loading, and 10X measurements on each sample with statistical rejection of outliers. The DR6000 also uses RFID technology to verify the expiry date of TNTplus test kits.Your water and environmental testing needs, all in one spectrophotometer.
Catalog No. 50-199-7797 Supplier Hach Company Supplier No. LPV441.99.00012
Add to Cart
Add to Cart
Request a Quote
The DR6000™ is the industry’s most advanced laboratory spectrophotometer for water testing. It offers high-speed wavelength scanning across the UV and Visible Spectra, and comes with over 250 pre-programmed methods including the most common water and environmental testing methods. With optional accessories allowing for high-volume testing via a carousel sample changer, and increased accuracy with a sample delivery system that eliminates optical difference errors, this instrument ensures you are ready to handle your wide-ranging water testing needs. Combining the DR6000 with Hach TNTplus™ test kits provides additional accuracy with guided step-by-step testing procedures, auto-method loading, and 10X measurements on each sample with statistical rejection of outliers. The DR6000 also uses RFID technology to verify the expiry date of TNTplus test kits.Your water and environmental testing needs, all in one spectrophotometer.