Learn More
Hach Company DR/Check Absorbance Standard Kit (Set of 4)
Supplier:  Hach Company 2763900
Encompass
Set contains four vials filled with gels at increasing Absorbance values. Used to confirm consistent instrument response.
Catalog No. 50-199-6642