Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ X-plore™ 6570 / Panorama Nova Mask Accessory, Overlay Protector

Catalog No. 19815805 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-815-805 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-815-805 Supplier Dräger™ Supplier No. 4055092
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clear overlay lens protector is made for the X-plore 6570 and Panorama Nova full facepiece.

  • Accessory for the X-plore 6570 ad Panorama full facepiece
  • Clear, self-adhesive lens protectors
  • Sold in 25/pack

Specifications

Color Clear
Disposable Single-use
Product Type Lens Protector
For Use With (Equipment) X-plore 6750 Mask, Panorama Nova Mask
Model 4055092
Quantity 25/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.