Learn More
Dräger™ Civil Defense Training Sets
Catalog No. 19822155 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Product Type:
Chlorine Tablets (CDS Set V)
Muriatic Acid (CDS Set I)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-822-155 Chlorine Tablets (CDS Set V)
19-822-154 Muriatic Acid (CDS Set I)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
These tubes are for Training ONLY.
Catalog No. 19-822-155 Supplier Dräger™ Supplier No. 8103240
Add to Cart
Add to Cart
These tubes are for Training ONLY.
For use with Civil Defense Simultest Set I and Set V
  • Cantain five tubes set mimics the tubes in CDS Set I and Set V
  • Muriatic acid (CDS Set I) and Chlorine tablets (CDS Set V) can be used to show color change
Specifications
Product Type Chlorine Tablets (CDS Set V)
For Use With (Equipment) Civil Defense Simultest Training Set
Includes 5-Tube Set, mimics the tubes in CDS set V