Learn More
Dräger™ P100 Cartridge for X-Plore 6000 and CDR 4500
Catalog No. 19500220 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Certifications/Compliance:
NIOSH
Catalog No. Certifications/Compliance
19-500-220 NIOSH
1 options
Catalog No. 19-500-220 Supplier Dräger™ Supplier No. 4055497
Specifications
Certifications/Compliance NIOSH
Connection Type 40 mm thread
Filter Type P100
Product Type Filter
For Use With (Application) For fit testing, to protect against all particulate aerosols, even those with oils present
For Use With (Equipment) Draeger fit test adapters
Quantity 1 Ea.