Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Panorama Nova Facepiece

Catalog No. 19057777 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-057-777 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-057-777 Supplier Dräger™ Supplier No. 4052955
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Highly reliable and secure full-face mask ideal for firefighting and mining.

  • Panorama Nova P EPDM - NFPA positive pressure mask
  • Protects during rescue activities and industrial workers
  • With various connection piece options
  • Compatible with SCBAs, CCBAs, respiratory filters and supplied airline systems
  • Without HUD

Specifications

Color Black
Product Type Full Facepiece Respirator
For Use With (Application) Wide range of applications
For Use With (Equipment) SCBA, supplied airline system
Harness Style Adjustable
Material EPDM
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.