Learn More
Dräger™ Panorama Nova RA Full-Facepiece
Catalog No. 19500224 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Includes:
Centrally located filter
Catalog No. Includes
19-500-224 Centrally located filter
1 options
This item is not returnable. View return policy
Catalog No. 19-500-224 Supplier Dräger™ Supplier No. R51535
This item is not returnable. View return policy
Classic, full-facepiece mask features triple sealing surfaces for a close, secure and comfortable fit without added pressure. Universal size fits all face types and eliminates the need for different size facepieces. An adjustable five-point rubber head strap aids in quick donning and doffing, making it ideal for use with SCBAs.
Features:
•     Shock-resistant polycarbonate lens provides a wide field of vision.
•     Elaborate flushing system prevents field of vision from being compromised and reduces fogging
•     Stainless-steel speech diaphragm ensures clear, reliable oral communication
•     Robust synthetic connector has both inhalation and exhalation valves
•     RA-type thread connection conforms to European standard EN 148-1 and accommodates a wide variety of Dräger respiratory filters
Order Info
  • Weight: 1.2 lb. (0.6kg)
  • Contact your Fisher Safety Customer Service Representative at 1-800-772-6733 for information about Dräger respiratory filters to fit your application.
Compliance
NIOSH approved
Both facepieces meet EN 136 standards
Specifications
Certifications/Compliance NIOSH, EN 136
Material Silicone, Polycarbonate Lens
Product Type Full Facepiece
Includes Centrally located filter
Suspension Type 5-point
Weight (English) 1.2 to 1.4 lb.
Weight (Metric) 0.55 to 0.65 kg