Learn More
Dräger™ SCBA Safety Mask
Catalog No. 19059914 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Panorama Nova; SCBA Face Mask
Catalog No.
19-059-914 Panorama Nova; SCBA Face Mask
1 options
Catalog No. 19-059-914 Supplier Dräger™ Supplier No. R53070
Inner mask and harness made of high quality silicone
  • Panorama Nova Silicone P PC facepiece
  • Without neck strap
Certifications
CE marked
Specifications
Certifications/Compliance EN 136
Connection Type Push-in
Product Type Facepiece
For Use With (Application) Supplies quickly and safely vital breathing air if the oxygen content drops below 17% by volume
For Use With (Equipment) Compressed air breathing apparatus
Harness Style Silicone
Material Silicone
Size Medium