Learn More
Dräger™ X-plore™ Series and Panorama Facepiece Accessory, Spectacle Kit
Catalog No. 19822152 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
X-plore and Panorama Facepieces
Catalog No. For Use With (Equipment)
19-822-152 X-plore and Panorama Facepieces
1 options
Catalog No. 19-822-152 Supplier Dräger™ Supplier No. 4056406
Add to Cart
Add to Cart
Spectacle kit accessory for use with X-plore series and Panorama facepieces.
  • Kit of lenses with plastic frame
Specifications
Material Plastic
Product Type Kit
For Use With (Application) To enhance field of vision, to provide maximum durability and comfort
For Use With (Equipment) X-plore and Panorama Facepieces
Includes Frame that can be fitted with prescription lens
Model 4056406