Learn More
Dräger™ Panorama Facepiece Rapid Adaptor
Catalog No. 19999274 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Disposable:
Reusable
Catalog No. Disposable
19-999-274 Reusable
1 options
Catalog No. 19-999-274 Supplier Dräger™ Supplier No. 4055251
Add to Cart
Add to Cart
Rapid adaptor enables conversion ability for the Panorama facepiece.
  • Converts Panorama Nova Mask to Air Purifying Respirator Mask
  • Converts Panorama Nova Mask for Fit Testing of Panorama Nova as Air Purifying Respirator Mask
  • Converts Panorama Nova Positive Pressure Mask to Negative Pressure Mask
Specifications
Disposable Reusable
Product Type Facepiece Adapter
For Use With (Equipment) Panorama Facepiece
Model 4055251