Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ X-plore™ Ammonia/Methylamine (AM/MA) Cartridge

Catalog No. 19985720 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Cartridge Type:
Ammonia, Methylamine, P100 Cartridge
Gas/Vapor Cartridge
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type
19-985-720 Ammonia, Methylamine, P100 Cartridge
17-985-670 Gas/Vapor Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-985-720 Supplier Dräger™ Supplier No. 6738037
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For protection against Ammonia and Methylamine gases

Fits Dräger X-plore 3300/3500 half masks and Dräger X-plore 5500 full face masks

  • Strong blue ABS plastic housing
  • Safe, quick and easy bayonet connection with clear guidance bringing the filter/cartridge securely into place
  • Both filters are inserted in a top-down position
  • Option to upgrade a gas cartridge to a combination cartridge by simply adding a pad retrainer and a N 95 or R 95 pad
  • Shelflife (from date of production): gas and combination cartridges 6 years, pads 3 years

  • To convert the AM/MA cartridge to a combination cartridge, you will need two filter pad retainers (p/n: 6738038) and two N95 Prefilters (p/n: 6738038).

Order Info

Packaged in pairs for optimum protection of the unused filter. Instructions included with each pair.

Certifications

NIOSH 42 CFR 84

Specifications

Cartridge Type Ammonia, Methylamine, P100 Cartridge
Certifications/Compliance NIOSH
Color Blue
Material ABS Plastic
Product Type Cartridge
For Use With (Equipment) Mask
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.