Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR Instruments Deluxe Dissecting Kit
SDPSCI_ED

Catalog No. S06840
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S06840 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

California Proposition 65 WARNING: Cancer and Reproductive Harm

Catalog No. S06840 Supplier DR Instruments Supplier No. 10GS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

California Proposition 65 WARNING: Cancer and Reproductive Harm

The Deluxe Dissecting Kit is an ideal choice for teachers and university students.

  • Kit contains most stainless-steel instruments in a zippered vinyl case
  • All tools are made with stainless-steel unless otherwise noted
  • Kit comes with hemostatic forceps

Includes:

  • Dissecting Iris scissors 4.5 in.
  • Dissecting scissor 5.5 in. with one point sharp and other point blunt
  • Dissecting forceps medium point 4.5 in. with guide pin
  • Tissue forceps 1 x 2
  • Dissecting teasing needles (one angular and one strraight) with metal chuck
  • Cartilage knife with 2 in. blade
  • Dissecting scalpel handle no. 3
  • Dissecting mall probe chrome
  • No. 10 blade or No. 15 blade
  • Vinyl zippered case with linen (black or red)

Specifications

Product Type Deluxe Dissecting Kit
Material Stainless Steel
For Use With (Application) An excellent choice for teachers and university students
Includes 4.5 in. dissecting iris scissors, 5.5 in. dissecting scissors with one point sharp and other point blunt, 4.5 in. dissecting forceps with guide pin, 1 x 2 tissue forceps, dissecting teasing needles (one angular and one strraight) with metal chuck
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.