Learn More
DR Instruments Dissecting Forceps with Curved Fine Points, Guide Pin, 4.5 in.
SDPSCI_ED
Catalog No. S08098
Change view
Click to view available options
Tip Style:
Curved Fine Points
Catalog No. Tip Style
S08098 Curved Fine Points
1 options
Catalog No. S08098 Supplier DR Instruments Supplier No. 16
Add to Cart
Add to Cart
Forceps for purposes involving delicate dissection.
  • Crafted with surgical grade stainless-steel
  • Guide pin for matching of points
  • Ridges for easy grip
  • Curved tips for flexibility to access hard-to-reach areas during dissection
  • Length: 11.43 cm (4.5 in.)
Specifications
Product Type Dissecting Forceps
Material Stainless Steel (Surgical Grade)
Includes Guide Pin
Tip Style Curved Fine Points
Length (English) 4.5 in.
Length (Metric) 11.4 cm