Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR Instruments Dissecting Forceps, Economy, 4.5 in.
SDPSCI_ED

Catalog No. S08099
Change view
Click to view available options
Tip Style:
Serrated Tips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
S08099 Serrated Tips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S08099 Supplier DR Instruments Supplier No. 17C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Forceps for starters used in various dissection experiments.

  • Made of chrome plated steel
  • Serrated tips and knurled handles for better grip in wet and dry use
  • Length: 11.43 cm (4.5 in.)

Specifications

Product Type Dissecting Forceps
Material Chrome-plated Steel
Tip Style Serrated Tips
Length (English) 4.5 in.
Length (Metric) 11.4 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.