Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR Instruments Dissecting Kit, Intermediate III
SDPSCI_ED

Catalog No. S06848
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S06848 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

California Proposition 65 WARNING: Cancer and Reproductive Harm

Catalog No. S06848 Supplier DR Instruments Supplier No. KIT2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

California Proposition 65 WARNING: Cancer and Reproductive Harm

Intermediate III Dissecting Kit is an ideal choice for teachers and students.

Ideal dissection kit for high school and early college classes. All dissection instruments are made with surgical stainless steel otherwise noted.

Includes:

  • Dissecting scissors 5.5 in. with one point sharp and other point blunt
  • Dissecting forceps 5 in.
  • Dissecting forceps SS, 4.5 in. with curved medium points
  • Two dissecting teasing needles (angular and straight) with plastic handle
  • Scalpel handle no. 4
  • Scalpel blade no. 24 (1 Piece)
  • Plastic ruler 6 in. with inches and centimeters markings
  • Glass dropper
  • Single fold vinyl case

Specifications

Product Type Intermediate III Dissecting Kit
For Use With (Application) Excellent choice for teachers and students
Includes Dissecting Scissors, 2 Dissecting Forceps, 2 Dissecting Teasing Needles, Scalpel Handle No. 4 And 24, Ruler, Glass Dropper, Single Fold Vinyl Case
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.