Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR Instruments Medium Points Forceps with Guide Pin, 4.5 in.
SDPSCI_ED

Catalog No. S08095
Change view
Click to view available options
Tip Style:
Serrated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
S08095 Serrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08095 Supplier DR Instruments Supplier No. 13
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Forceps made with 410-Stainless-Steel, medium points for dissecting purposes.

  • Features serrated points
  • Guide pin for matching of points
  • Ridges for easy grip
  • Length: 11.43 cm (4.5 in.)

Specifications

Material Stainless Steel (410)
Includes Guide Pin
Tip Style Serrated
Tip Thickness Medium
Length (English) 4.5 in.
Length (Metric) 11.4 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.