Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR Instruments Angular Probe and Seeker, Stainless steel, 6 in.
SDPSCI_ED

Catalog No. S08110
Change view
Click to view available options
Tip Style:
Angular Probe, Straight Seeker
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
S08110 Angular Probe, Straight Seeker
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08110 Supplier DR Instruments Supplier No. 35ANG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Angular Probe and Seeker used when working with vessels, nerves, and muscles.

  • Made from durable stainless steel
  • Features a sharp point at one end and an angular medium point on opposite side
  • Length: 15.24 cm (6 in.)

Specifications

Product Type Probe
Tip Style Angular Probe, Straight Seeker
Length (English) 6 in.
Length (Metric) 15.24 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.