Learn More
DR Instruments Scalpel Handles, Stainless Steel
SDPSCI_ED
Catalog No. p-4789158
Change view
Click to view available options
Size:
#3
#4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S08115 #3
S08116 #4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08115 Supplier DR Instruments Supplier No. 27
Only null left
Add to Cart
Edge
Add to Cart
Stainless Steel Scalpel Handles for finer dissection in various surgical purposes. Uses multiple size blades to fit your dissection needs.
  • Made with Stainless-steel
  • Slip lock design
Specifications
Blade Material Stainless Steel
Product Type Scalpel Handle
Size #3
For Use With (Equipment) Blade: # 10, # 11, # 12 and # 15
Description Slip Lock