Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR Instruments Student Scalpel, Stainless Steel, 1.5 in. Super Sharp Blade
SDPSCI_ED

Catalog No. S08111
Change view
Click to view available options
Length (Metric) Blade:
3.8 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Blade
S08111 3.8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08111 Supplier DR Instruments Supplier No. 23
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Student Scalpel made with fine quality stainless steel for dissection purposes.

  • One piece scalpel with 1.5 in. super sharp blade.
  • Blade length: 3.8 cm (1.5 in.)
  • Fine quality stainless steel.
  • Hand honed super sharp blade.
  • Ideal for all your dissection labs.
  • Ideal for cutting preserved specimens.

Specifications

Blade Material Chrome-plated Steel
Product Type Scalpel
Length (Metric) Blade 3.8 cm
Length (English) 1.5 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.