Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR Instruments 1 x 2 Teeth Tissue Forceps or Rat Tooth Tissue Forceps 5.5 in., Stainless Steel
SDPSCI_ED

Catalog No. S08100
Change view
Click to view available options
Tip Style:
Rat Tooth
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
S08100 Rat Tooth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08100 Supplier DR Instruments Supplier No. 12T
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Forceps Specially designed teeth for holding tissues or skin during dissection.

  • Made with Stainless-steel
  • 1 x 2 teeth points for holding tissues and skin
  • Serrated handles for non-slip grip
  • Length: 14 cm (5.5 in.)
  • 1 x 2 teeth

Specifications

Product Type Tissue Forceps
Material Stainless Steel
Teeth Count 1x2
Tip Style Rat Tooth
Length (English) 5.5 in.
Length (Metric) 12 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.