Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR Instruments Veterinary Dissection Kit for First Year Vet-Students
SDPSCI_ED

Catalog No. S07893
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07893 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

California Proposition 65 WARNING: Cancer and Reproductive Harm

Catalog No. S07893 Supplier DR Instruments Supplier No. DRVT3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

California Proposition 65 WARNING: Cancer and Reproductive Harm

Contains all necessary instruments required for learning anatomy of animals.

Veterinary dissection kit is a valuable educational tool that provides hands-on to study about the anatomy of various animals, including domestic and wildlife preserved specimens. This veterinary student training kit contains the most basic tools that are required to learn anatomy of the most preserved specimens. DR Instruments veterinary dissection kit allows students to explore the structure and function of various organs, tissues, and systems, which is essential for understanding animal biology, physiology, and pathology, in a classroom or laboratory environment.

Includes:

  • Scalpel Handle No. 4
  • Scalpel Blades No. 20 and No. 22 (10 pieces each)
  • Tissue Forceps with 2 x 3 Teeth
  • Addison Forceps with 1 x 2 Teeth
  • Thumb Dressing Forceps, 5.5 in. (140 mm)
  • Hemostatic Forceps (Kelly or Criel)
  • Dissection Scissors 6 in. (152 mm), Sharp/Sharp (Two sharp point blades)
  • Iris Scissors, 4.5 in. (114.3 mm)
  • Huber Probe with one end blunt and bent, and other end chisel
  • Deluxe Dissection Kit Case to hold all tools

Specifications

Product Type Veterinary Student Dissecting Kit
For Use With (Application) For First year gross anatomy class
Includes Scalpel Handle, Scalpel Blades, Tissue Forceps, Addison Forceps, Thumb Dressing Forceps, Hemostatic Forceps, Dissection Scissors, Iris Scissors, Huber Probe, Case
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.